Learn More
Selleck Chemical LLC SIS3 HCl 2mg 521984-48-5
SDP
Supplier:  Selleck Chemical LLC S79592MG
Encompass_Preferred
SIS3, a novel specific inhibitor of Smad3, inhibits TGF-? and activin signaling by suppressing Smad3 phosphorylation without affecting the MAPK/p38, ERK, or PI3-kinase signaling pathways. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5480