Learn More
Medchemexpress LLC Sisomicin (sulfate) | 53179-09-2 | 99.3% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB122210MM/1ML
Encompass_Preferred
Sisomicin sulfate is a broad-spectrum aminoglycoside antibiotic produced by *Micromonospora inyoensis*. It is highly active against Gram-positive bacteria. It completely inhibits the growth of various clinical isolates of bacteria, such as *Klebsiella*, *Salmonella*, *Citrobacter*, and *Staphylococcus aureus* at 6.25 μg/mL.
  • Broad-spectrum aminoglycoside antibiotic.
  • Highly active against Gram-positive bacteria.
  • Completely inhibits the growth of various clinical isolates bacteria.
Catalog No. 50-004-12851