Learn More
Cayman Chemical SIsopropyl IsothIoureahydr
SDP
Supplier:  Cayman Chemical 81290500 MG
Encompass_Preferred
IPTU is a potent inhibitor of NOS in vitro but may lack good in vivo efficacy due to poor cellular penetration. The Ki values are 9.8, 22, and 37 nM using purified human iNOS, eNOS, and nNOS, respectively.{1292}
Catalog No. 50-289-5419