Learn More
Abcam Sivelestat, neutrophil elastase inhibitor, 5MG
Supplier:  Abcam AB1461845MG
Encompass_Preferred
MW 434.5 Da. Potent, selective, substrate competitive neutrophil elastase inhibitor (IC₅₀ values are 19-49 nM and 5.6 μM for leukocyte and pancreas elastase respectively). Suppresses neutrophil-induced capillary permeability and leukocyte kinetics. Active in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4869