Learn More
Apexbio Technology LLC Sivelestat sodium tetrahydrate, 100mg CAS# 201677-61-4
SDP
Supplier:  Apexbio Technology LLC B8201100
Encompass_Preferred
Sivelestat sodium tetrahydrate is a potent competitive inhibitor of human neutrophil elastase (HNE) with IC50 of 44 nM and Ki of 0.2 M. It is also known as ONO5046, LY544349 and EI546. It can also inhibit leukocyte elastase in rabbit, rat, hamster and mouse. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8251