Learn More
Selleck Chemical LLC Sivelestat sodium tetrahydrate S4666-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S466610MM/1ML
Encompass_Preferred
Sivelestat sodium tetrahydrate (ONO5046-Na EI546 LY544349) is a competitive inhibitor of human neutrophil elastase also inhibits leukocyte elastase obtained from rabbit rat hamster and mouse
Catalog No. 50-310-8632