Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Premier Protein SEC Column, 250Å, 2.5 μm, 7.8 x 300 mm

Supplier:  Waters Corp WTC030S5

Encompass_Preferred

The XBridge Premier Protein SEC Column is specifically designed and optimized for the analysis of proteins using Size Exclusion Chromatography with Multi-Angle Light Scattering (SEC-MALS) detection. This column ensures high-resolution separations for accurate protein characterization.

  • Optimized for protein analysis by SEC-MALS.
  • Features a bead size of 2.5 μm for efficient separations.
  • Provides a pore size of 250 Å suitable for protein applications.
  • Measures 7.8 mm internal diameter by 300 mm in length.

Catalog No. NC9960343


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.