Learn More
Medchemexpress LLC SJ3149 CK1α Molecular Glue Degrader | 3026986-17-1 | 99.48% | 376.37 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16044410MM/1ML
Encompass_Preferred
SJ3149 is a selective and potent molecular glue degrader of CK1α protein with broad antiproliferative activity, intended for use in cancer research. It effectively degrades CK1α protein in MOLM-13 cells at low nanomolar concentrations and inhibits cell growth. In vivo studies also show significant degradation of CK1α protein levels in mice.
  • Selective and potent molecular glue degrader of CK1α protein.
  • Broad antiproliferative activity against cancer cells.
  • Degrades CK1α protein in MOLM-13 cells at 4 nM.
  • Inhibits MOLM-13 cell growth with an IC50 of 14 nM.
  • Significantly degrades CK1α protein levels in NSG mice at 50 mg/kg.
Catalog No. 50-003-91409