Learn More
Medchemexpress LLC SJ6986, Solution 10 mM * 1 mL in DMSO | CAS No. : 2765625-93-0
SDP
Supplier:  Medchemexpress LLC HY13219910MM 1ML/DMSO
Encompass_Preferred
SJ6986 is a potent, selective and orally active GSPT1/2 Molecular Glue degrader, with a DC50 of 2.1 nM (Dmax 99%) for GSPT1.
Catalog No. NC3914007