Learn More
Medchemexpress LLC HY-19556 5mg Medchemexpress, (+)-SJ733 CAS:1424799-20-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY195565MG
Encompass_Preferred
Medchemexpress, HY-19556 5mg (+)-SJ733 CAS:1424799-20-1 (+)-SJ733 is an anti-malaria agent which can also inhibit Na+-ATPase PfATP4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2144