Learn More
Medchemexpress LLC SJPYT-195 | 2973762-16-0 | C35H34N6O8 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY150616
Encompass_Preferred
SJPYT-195 is a cytotoxic GSPT1 degrader that can be utilized for PROTAC synthesis. It effectively reduces endogenous PXR protein in colorectal SNU-C4 cell lines by degrading GSPT1. This compound is supplied as a white to off-white solid, with a purity of 98.07%, and is intended for research use only.
  • Cytotoxic GSPT1 degrader
  • Utilized for PROTAC synthesis
  • Potently reduces endogenous PXR protein
  • Mechanism involves GSPT1 degradation
  • Supplied as a white to off-white solid
  • High purity (98.07%)
  • For research use only
Catalog No. 50-003-74303