Learn More
Medchemexpress LLC HY-137341 5mg , SK-3-91 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1373415MG
Encompass_Preferred
HY-137341 5mg SK-3-91 CAS: SK-3-91 is a multi-kinase degrader. SK-3-91 could collectively induce degradation of the largest number of unique kinases (>125 unique kinases). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0093