Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SK1-I (BML-258) | 1072443-89-0 | 98.8% | 277.40 | C17H27NO2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1190165MG

Encompass_Preferred

SK1-I (BML-258) is a sphingosine analog and an isozyme-specific competitive inhibitor of sphingosine kinase 1 (SPHK1) with a reported Ki of about 10 μM. It enhances autophagy and has demonstrated antitumor activity. Intended for research use only.

  • Isozyme-specific SPHK1 inhibition (Ki ≈ 10 μM).
  • Sphingosine analog scaffold.
  • High purity: 98.83%.
  • Molecular weight 277.40 g/mol; formula C17H27NO2.
  • Available as small solid quantities and as a 10 mM solution in DMSO.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-29201


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.