Learn More
Medchemexpress LLC HY-101805 1mg Medchemexpress, SK1-IN-1 CAS:1218816-71-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1018051MG
Encompass_Preferred
Medchemexpress, HY-101805 1mg SK1-IN-1 CAS:1218816-71-7 SK1-IN-1 is a potent sphingosine kinase 1 (SPHK1) inhibitor with an IC50 of 58 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7875
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.