Learn More
Medchemexpress LLC HY-111655 10mg Medchemexpress, SKA-31 CAS:40172-65-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11165510MG
Encompass_Preferred
Medchemexpress, HY-111655 10mg SKA-31 CAS:40172-65-4 SKA-31 is a potent potassium channel activator with EC50s of 260 nM, 1.9 μM, 2.9 μM, and 2.9 μM for KCa3.1, KCa2.2, KCa2.1 and KCa2.3, respectively. SKA-31 potentiates endothelium-derived hyperpolarizing factor response and lowers blood pressure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8939