Learn More
ATCC SK-BR-3; Breast Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB30
Encompass
SK-BR-3 [SKBR3] cells were isolated in 1970 from pleural effusion cells of a 43-year-old, White, female adenocarcinoma patient with blood type A+ who had been treated with radiation, steroids, cytoxan, and 5-fluorouracil. Use these cells in your breast cancer research.
Catalog No. 50-238-2871
 Restricted Item