Learn More
Jeio Tech Inc Jeio Tech™ SKC-7100, Programmable Large Capacity Open Air Orbital Shaker (120V, 60Hz, US Plug)
SDP
Supplier:  Jeio Tech Inc AAH30216U
Encompass_Preferred
Jeio Tech SKC Programmable Shakers are high-performance orbital shakers designed for precise and repeatable mixing applications. Equipped with an intuitive programmable controller, the SKC series allows users to set multi-step shaking profiles with customizable speed, time, and pause functions. Ideal for complex workflows in molecular biology, biochemistry, and pharmaceutical labs, these shakers provide smooth and stable motion across a wide speed range. Their durable construction, quiet operation, and versatile platform compatibility make them an excellent choice for demanding laboratory environments requiring reliable and automated shaking performance. Shaking platform required; sold separately.
Catalog No. NC3062464
Only null left
Add to Cart
Request a Quote