Learn More
Medchemexpress LLC Skeletal muscle-targeted peptide MSP | 216763-24-5 | 98.7% | 674.74 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP10643
Encompass_Preferred
Skeletal muscle-targeted peptide MSP is a 7-amino-acid (ASSLNIA) muscle-targeting peptide (MTP). It is capable of targeting viruses through various binding ligands to the muscle and can be utilized for research into diseases affecting the heart and skeletal muscles.
  • 7-amino-acid (ASSLNIA) muscle-targeting peptide (MTP)
  • Capable of targeting viruses through various binding ligands to the muscle
  • Utilized for research into diseases affecting the heart and skeletal muscles
Catalog No. 50-004-26653