Learn More
Medchemexpress LLC Skepinone-L | 1221485-83-1 | MFCD22572738 | C24H21F2NO4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY153001MG
Encompass_Preferred
Skepinone-L is a chemical probe and a selective p38 mitogen-activated protein kinase inhibitor. It is the first ATP-competitive p38 MAPK inhibitor with excellent in vivo efficacy and selectivity, making it a valuable probe for chemical biology research and potentially fostering the development of a unique class of kinase inhibitors.
  • Selective p38 mitogen-activated protein kinase inhibitor
  • First ATP-competitive p38 MAPK inhibitor
  • Excellent in vivo efficacy and selectivity
  • Valuable probe for chemical biology research
  • Potentially fosters the development of a unique class of kinase inhibitors
Catalog No. 50-003-72517