Learn More
ATCC SK-ES-1; Ewing's Sarcoma; Human (Homo sapiens)
Supplier:  ATCC HTB86
Encompass
SK-ES-1 is a cell line exhibiting epithelial morphology that was isolated from the bones of an 18-year-old, White male patient with anaplastic osteosarcoma. This product has applications in cancer research.
Catalog No. 50-238-4020