Learn More
Medchemexpress LLC SKF-34288 hydrochloride | 320386-54-7 | 98.6% | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY12892310MM/1ML
Encompass_Preferred
SKF-34288 (3-Mercaptopicolinic acid) hydrochloride is an orally active phosphoenolpyruvate carboxykinase (PEPCK) inhibitor with a Ki of 2-9 μM. It acts as a potent hypoglycemic agent by inhibiting glucose synthesis. Additionally, it inhibits Asn metabolism and leads to an increase in amino acids and amides. It is intended for research use only.
  • Potent phosphoenolpyruvate carboxykinase (PEPCK) inhibitor
  • Inhibits glucose synthesis
  • Increases amino acids and amides by inhibiting Asn metabolism
  • Inhibits C2C12 cell proliferation
  • Induces myogenic differentiation of C2C12 cells
  • Reduces mRNA expression of Pck2 and genes associated with serine biosynthesis
  • Reduces blood glucose in starved rats
  • Neutralizes Salbutamol-mediated hyperglycaemia in rabbits
  • Reduces pyruvate-induced blood glucose level in rats
Catalog No. 50-003-38930