Learn More
Medchemexpress LLC SKF 38393 hydrochloride | 62717-42-4 | MFCD00069248 | 99.5% | 291.77 g/mol | C16H18ClNO2 | 50MG

Supplier: Medchemexpress LLC HY12520A50MG
SKF 38393 hydrochloride is a research-grade selective agonist of the dopamine D1 receptor with a reported IC50 of 110 nM. Supplied as a hydrochloride salt with high analytical purity, it is intended for laboratory research and analytical applications; not for human or veterinary use.
- Selective D1 receptor agonist with IC50 = 110 nM.
- Hydrochloride salt; molecular formula C16H18ClNO2; molecular weight 291.77 g/mol.
- Purity approximately 99.5% by HPLC.
- Available in small-scale solid quantities and ready-to-use solution formats for experiments.
- Store solid sealed at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
- For research use only; follow the safety datasheet and handling instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.