Learn More
Cayman Chemical SKF 38393hydrobromIde 10mg
SDP
Supplier:  Cayman Chemical 1915010 MG
Encompass_Preferred
A partial agonist of the dopamine D1-like receptors D1 and D5 (Kis = 1 and ~0.5 nM, respectively)
Catalog No. 50-298-3293