Learn More
Medchemexpress LLC Skf-75670 hydrobromide | 62717-63-9 | 99.8% | 350.25 | C17H20BrNO2 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1250435MG
Encompass_Preferred
SKF-75670 hydrobromide is a research chemical that functions as a dopamine D1 receptor partial agonist and has been reported to act as a cocaine antagonist. It is supplied as the hydrobromide salt in solid form and as a DMSO solution for use in in vitro and in vivo pharmacology studies.
  • Dopamine D1 receptor partial agonist and cocaine antagonist.
  • Hydrobromide salt supplied as solid and 10 mM DMSO solution.
  • High purity (99.8%) suitable for in vitro and in vivo studies.
  • Available in small research pack sizes, including 5 mg.
  • CAS 62717-63-9, chemical formula C17H20BrNO2, molecular weight 350.25.
Catalog No. 50-002-35294