Learn More
Cayman Chemical -SKF 81297hydrobromIde 10mg
SDP
Supplier:  Cayman Chemical 1506710 MG
Encompass_Preferred
A selective agonist of the dopamine D1-like receptor (Ki = 1.9 nM) that demonstrates comparatively lower binding affinity for the dopamine D2, dopamine D3, serotonin 5-HT2A, and adrenergic α2 receptors (Kis = 1,272, >10,000, 955, and 509 nM, respectively)
Catalog No. 50-297-0112