Learn More
Medchemexpress LLC SKF-83566 | 99295-33-7 | 99.8% | 332.23 | C17H18BrNO | 5 MG
SDP
Supplier:  Medchemexpress LLC HY103430A5MG
Encompass_Preferred
SKF-83566 is a potent, blood-brain permeable, and orally active D1-like dopamine receptor antagonist (CAS 99295-33-7) used in pharmacological research. It also displays weaker competitive antagonism at 5-HT2 receptors, inhibits dopamine transporter activity in some assays, and can selectively inhibit adenylyl cyclase 2.
  • Potent D1-like dopamine receptor antagonist suitable for receptor pharmacology studies.
  • Blood-brain permeable and orally active in preclinical models.
  • High purity solid (~99.8%) for consistent experimental results.
  • Soluble in DMSO at approximately 33.33 mg/mL; ultrasonication may be required.
  • Stable when stored as a powder at -20°C for long-term storage.
  • Commonly used in Parkinson's disease and nicotine craving research.
Catalog No. 50-002-46808