Learn More
Medchemexpress LLC SKF-96365 hydrochlor 10mg
SDP
Supplier:  Medchemexpress LLC HY10000110MG
Encompass_Preferred
SKF-96365 hydrochloride is a potent TRP channel blocker and a store-operated Ca2 entry (SOCE) inhibitor SKF-96365 hydrochloride significantly inhibits hERG hKCNQ1/hKCNE1 hKir2 1 and hKv4 3 current and significantly prolongs the QTc interval in isolated guinea pig hearts SKF-96365 hydrochloride exhibits potent anti-neoplastic activity by inducing cell-cycle arrest and apoptosis in colorectal cancer cells1]2]
Catalog No. 50-001-68922