Learn More
Cayman Chemical SKF 96365 hydrochloride
SDP
Supplier:  Cayman Chemical 1000931250 MG
Encompass_Preferred
Inhibits the receptor-mediated influx of calcium via voltage-gated calcium channels (IC50 = 10 µM); inhibits the acetylcholine-induced depolarization of circular smooth muscle in a dose-dependent manner at 3-50 µM
Catalog No. 50-289-0879