Learn More
Abcam SKI-I, Sphingosine Kinase Inhibitor, 25MG
Supplier:  Abcam AB14220925MG
Encompass_Preferred
MW 410.5 Da, Purity >90%. Specific non-lipid sphingosine kinase (SK) inhibitor. Inhibits SK1 and SK2. Inhibits sphingosine 1-phosphate (S1P) production and pro-apoptotic. Induces autophagy. Cell permeable and active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6282