Learn More
Selleck Chemical LLC SKI II 25mg 312636-16-1 SphK-I2
SDP
Supplier:  Selleck Chemical LLC S717625MG
Encompass_Preferred
SKI II (SphK-I2) is a highly selective and non ATP-competitive¿sphingosine kinase (SphK)¿inhibitor with¿IC50¿of 0.5 ?M, while exhibiting no inhibitory action on other kinases including PI3K, PKC? and ERK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4488