Learn More
Enterprise Technology Solutions SKILCRAFT Gloves,Exam,Nitrile,Textured,PF,Large,100/BX,Blue
SDP
Supplier:  Enterprise Technology Solutions NSNNIB0313
Encompass_Preferred
Chemo-Tested Exam Gloves are tested for use with chemotherapy drugs in accordance with ASTM 6978. Made of nitrile, they are powder-free, latex-free, odor-free and contain no natural rubber proteins. These disposable gloves with a textured grip are easy to put on. They are puncture-resistant and chemical-resistant so the material is superior to latex and vinyl. Gloves are mandatory only for U.S. Department of Veterans Affairs.
Catalog No. 50-008-28324