Learn More
Medchemexpress LLC Skimmin (Umbelliferone glucoside) | 93-39-0 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN22631MG
Encompass_Preferred
Skimmin (Umbelliferone glucoside) is a pharmacologically active molecule derived from Hydrangea paniculata, known for its use in traditional Chinese medicine as an anti-inflammatory agent. It exhibits renal protective properties by improving creatinine clearance and reducing kidney injuries. Additionally, Skimmin demonstrates anti-amoebic, anti-cancer, and neuroprotective activities.
  • Oral bioavailability
  • Anti-inflammatory agent
  • Renal protective activity
  • Reduces plasma creatinine and kidney injuries
  • Anti-amoebic activity against Entamoeba histolytica
  • Anti-cancer activity
  • Neuroprotective activity
  • Reduces cardiac fibrosis
  • Decreases inflammatory markers in cardiac tissues
  • Potential for diabetes and related disease research
Catalog No. 50-004-17834