Learn More
Selleck Chemical LLC SKL2001 10mg 909089-13-0
SDP
Supplier:  Selleck Chemical LLC S832010MG
Encompass_Preferred
SKL2001 is a novel agonist of the Wnt/?-catenin pathway. It disrupts the Axin/?-catenin interaction. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5941