Learn More
Medchemexpress LLC Sklb325 | 66680-03-3 | 99.2% | C12H12N4O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13978210MG
Encompass_Preferred
SKLB325 is a small-molecule JMJD6 inhibitor supplied for research use. It is offered as a high-purity compound intended for in vitro and laboratory studies to probe JMJD6-related biology and epigenetic mechanisms.
  • High purity suitable for research applications.
  • Small-molecule inhibitor for in vitro studies.
  • Molecular formula C12H12N4O2 and molecular weight 244.25 g·mol-1.
  • Supplied with safety documentation for laboratory handling.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-58185