Learn More
ATCC SK-MEL-28; Melanoma; Human (Homo sapiens)
Supplier:  ATCC HTB72
Encompass
SK-MEL-28 cells are melanocytes isolated from the skin tissue of a 51-year-old, male patient with malignant melanoma. This cell line is a suitable transfection host and has applications in cancer and toxicology research. SK-MEL-28 is one of a very extensive series of melanoma cell lines isolated by T. Takahashi and associates.
Catalog No. 50-238-4233