Learn More
ACCEGEN BIOTECH SKN-3, a Head and Neck Cancer Cell Line, 1 Vial, 1.5 ml
Supplier:  ACCEGEN BIOTECH ABCTC1043
Encompass
SKN-3 is a head and neck cancer cell line carrying NSD1 loss of function mutation. This is only one of the few NSD1 mutated head and neck cancer cell line commercially available.
Catalog No. NC2831875