Learn More
Selleck Chemical LLC Skp2 inhibitor C1
SDP
Supplier:  Selleck Chemical LLC S8652100MG
Encompass_Preferred
Skp2 inhibitor C1 (SKPin C1) is a specific and selective small-molecule inhibitor of Skp2-mediated p27 degradation by reducing p27 binding through key compound-receptor contacts
Catalog No. 50-000-01172