Learn More
Apexbio Technology LLC SL-327 305350-87-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A18945.1
Encompass_Preferred
SL-327 (CAS 305350-87-2) is a selective inhibitor targeting MEK1 and MEK2 kinases exhibiting IC50 values of 0 18 M and 0 22 M respectively MEK1/2 mediate the phosphorylation and activation of extracellular signal-regulated kinases (ERK) crucial components of the MAPK/ERK signaling pathway SL-327 treatment lowers phosphorylated ERK levels and subsequent activation of transcription factors including CREB Elk-1 and c-Fos influencing gene expression linked to synaptic plasticity In animal models SL-327 modulates drug-induced behaviors and neuronal plasticity processes making it a relevant molecular tool to investigate MAPK/ERK signaling pathways in neuroscience research
Catalog No. 50-000-62795