Learn More
Selleck Chemical LLC SLC13A5-IN-1 S0755-25MG
SDP
Supplier:  Selleck Chemical LLC S075525MG
Encompass_Preferred
SLC13A5-IN-1 is a selective inhibitor of sodium-citrate co-transporter (SLC13A5/NaCT) It completely blocks the uptake of 14 C-citrate and has the potential for the treatment of metabolic and/or cardiovascular diseases
Catalog No. 50-000-42570