Learn More
ABclonal Technology SLC25A4/ANT1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A150271000UL
Encompass_Preferred
This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the cytoplasm into the mitochondrial matrix and ATP from the mitochondrial matrix into the cytoplasm. The protein forms a homodimer embedded in the inner mitochondria membrane. Mutations in this gene have been shown to result in autosomal dominant progressive external opthalmoplegia and familial hypertrophic cardiomyopathy.
Catalog No. 50-001-08303