Learn More
Aviva Systems Biology Corp SLC26A8 Antibody - C-terminal region (ARP44085_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44085_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC26A8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SLC26A8. Alternate names: RP11-482O9.1, FLJ32714, TAT1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-58711