Learn More
Medchemexpress LLC SLC5A1 protein (sodium/glucose cotransporter 1) | 50UG

Supplier: Medchemexpress LLC HYP70320750UG
Recombinant human SLC5A1 (sodium/glucose cotransporter 1, SGLT1) expressed in HEK293 cells with a His tag. Supplied as a 0.22 μm filtered solution formulated in 20 mM HEPES (pH 7.5), 150 mM NaCl, and trace detergents to stabilize the membrane protein. Molecular weight is ~75.8 kDa (M1-A664). Intended for biochemical and functional studies.
- Species: Human.
- Expression system: HEK293 cells.
- Tag: His tag.
- Accession: P13866-1.
- Amino acid range: M1-A664.
- Molecular weight: 75.8 kDa.
- Formulation: 20 mM HEPES pH 7.5, 150 mM NaCl, 0.001% (w/v) LMNG, 0.00033% (w/v) GDN, 0.00013% (w/v) CHS.
- Endotoxin: <1 EU/μg (LAL method).
- Storage and handling: store at -80°C, ship on dry ice, avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.