Learn More
ABclonal Technology SLC5A1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A119761000UL
Encompass_Preferred
This gene encodes a member of the sodium-dependent glucose transporter (SGLT) family. The encoded integral membrane protein is the primary mediator of dietary glucose and galactose uptake from the intestinal lumen. Mutations in this gene have been associated with glucose-galactose malabsorption. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-14853