Learn More
ABclonal Technology SLC7A1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A147841000UL
Encompass_Preferred
Enables L-arginine transmembrane transporter activity and L-histidine transmembrane transporter activity. Involved in amino acid transport. Located in membrane. Part of apical plasma membrane, basolateral plasma membrane, and protein-containing complex.
Catalog No. 50-001-06455