Learn More
Medchemexpress LLC SLF 10mM 1mL | 195513-96-3 | 524.65 | C30H40N2O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11487210MM 1ML
Encompass_Preferred
SLF is a synthetic ligand of FK506-binding proteins (FKBP) used as a ligand in targeted protein degradation and FKBP-targeted studies. It binds FKBP51 (affinity 3.1 μM) and inhibits FKBP12 (IC50 2.6 μM). Supplied as a concentrated solution for biochemical and in vivo use.
  • Synthetic ligand for FKBP proteins.
  • Useful for design and synthesis of PROTACs.
  • Affinity for FKBP51: 3.1 μM; IC50 for FKBP12: 2.6 μM.
  • Molecular formula C30H40N2O6; molecular weight 524.65.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High DMSO solubility (100 mg/mL) and demonstrated in vivo formulation solubility.
Catalog No. 50-003-07869