Learn More
Hampton Research HR2-070/Slice pH - kit contains 500 microliters of 96 reagents in a V-Bottom (conical bottom) 96 well polypropylene microplate, user guide, formulations and a sealing film
SDP
Supplier:  Hampton Research HR2070
Encompass
HR2-070/Slice pH - kit contains 500 microliters of 96 reagents in a V-Bottom (conical bottom) 96 well polypropylene microplate, user guide, formulations and a sealing film. Please note items from this supplier are non-returnable.
Catalog No. NC0156879