Learn More
Hampton Research SLICE PH REAGENT #05 (A5)
SDP
Supplier:  Hampton Research HR207005
Encompass
HR2-070-05/Slice pH #05 (A5), 185 ml - 1.0 M Sodium citrate tribasic dihydrate pH 3.6. Please note items from this supplier are non-returnable.
Catalog No. 50-562-863