Learn More
Hampton Research SLICE PH REAGENT #39 (D3)
SDP
Supplier:  Hampton Research HR207039
Encompass
HR2-070-39/Slice pH #39 (D3), 185 ml - 1.0 M ADA pH 5.8. Please note items from this supplier are non-returnable.
Catalog No. 50-562-897