Learn More
Hampton Research SLICE PH REAGENT #61 (F1)
SDP
Supplier:  Hampton Research HR207061
Encompass
HR2-070-61/Slice pH #61 (F1), 185 ml - 1.0 M HEPES sodium pH 7.5. Please note items from this supplier are non-returnable.
Catalog No. 50-562-919