Learn More
Sigma Aldrich Fine Chemicals Biosciences Slide box 100 place blue
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS15991A
Encompass
Standard Microscope Slide Boxes Constructed of durable polypropyleneSlide boxes hold either 25 or 100 standard slides (25 x 75 mm and 1 x 3 inches).Hinged lids are held securely closed with a latch lockStacking feet maximize stabilitySlide identification card - Capacity 100 slides Hinge Latch Lock 210 x 169 x 37 mm Blue
Catalog No. 50-290-1471